Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GDNF family receptor alpha 3 | GFRA3 | GDNFR3 | -29.9kb | Sense | Chr. 5 - 138252380 - 138274564 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 53 member C | FAM53C | C5orf6 | 29.5kb | Anti-sense | Chr. 5 - 138337535 - 138349729 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map