Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MHC class I polypeptide-related sequence A | MICA |
MIC-A, PERB11.1 |
-26.4kb | Sense | Chr. 6 - 31399784 - 31415315 on NCBI Build GRCh38 |
| Downstream | HLA complex P5 (non-protein coding) | HCP5 |
6S2650E, D6S2650E, P5-1 |
16.2kb | Sense | Chr. 6 - 31463180 - 31465809 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map