Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4324 | MIR4324 | mir-4324 | -14.0kb | Sense | Chr. 19 - 49308797 - 49308868 on NCBI Build GRCh38 |
| Downstream | CD37 molecule | CD37 |
GP52-40, TSPAN26 |
10.2kb | Anti-sense | Chr. 19 - 49335314 - 49340608 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map