Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peroxidasin like | PXDNL |
PMR1, PRM1, VPO2 |
Sense | Chr. 8 - 51319575 - 51811031 on NCBI Build GRCh38 | |
| Downstream | protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 | PCMTD1 | 71.0kb | Sense | Chr. 8 - 51817575 - 51899186 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map