Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | crystallin lambda 1 | CRYL1 |
GDH, HEL30, lambda-CRY |
-84.0kb | Anti-sense | Chr. 13 - 20403667 - 20525873 on NCBI Build GRCh38 |
| Downstream | interleukin 17D | IL17D | IL-17D | 92.2kb | Sense | Chr. 13 - 20702775 - 20723100 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map