| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000412773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000412773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000422876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000422876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000411511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008254.hg.1 |
ENST00000432495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000412773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000412773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000422876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000422876.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008269.hg.1 |
ENST00000453811.1 |
Ensembl |