Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arginyltransferase 1 | ATE1 | Anti-sense | Chr. 10 - 121740421 - 121928801 on NCBI Build GRCh38 | ||
| Downstream | ATE1 antisense RNA 1 | ATE1-AS1 | 77.2kb | Sense | Chr. 10 - 121928312 - 121951965 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map