Assay Details
| Gene Symbol: LOC101929297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125862.1 | 2 - 3 | 1193 | 85 | ||
| GenBank mRNA | BC040873.1 | - | 2 - 3 | 1193 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T brachyury transcription factor | T |
SAVA, TFT |
-74.1kb | Anti-sense | Chr. 6 - 166157644 - 166168669 on NCBI Build GRCh38 |
| Downstream | proline rich 18 | PRR18 | 51.2kb | Anti-sense | Chr. 6 - 166305680 - 166308383 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929297
Genomic Map