Assay Details
| Gene Symbol: LOC101928389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135631.1 | 1 - 2 | 322 | 107 | ||
| GenBank mRNA | BC040915.1 | - | 1 - 2 | 322 | 107 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM549840.1 | - | 321 | Chr. - - [-] on Build | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NHS antisense RNA 1 | NHS-AS1 | Anti-sense | Chr. X - 17552349 - 17559127 on NCBI Build GRCh38 | ||
| Downstream | sex comb on midleg-like 1 (Drosophila) | SCML1 | 150.3kb | Sense | Chr. X - 17737449 - 17754988 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928389
Genomic Map