Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RORA antisense RNA 1 | RORA-AS1 | Anti-sense | Chr. 15 - 60479178 - 60630639 on NCBI Build GRCh38 | ||
| Downstream | RORA antisense RNA 2 | RORA-AS2 | 100.2kb | Anti-sense | Chr. 15 - 60681569 - 60687249 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map