Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 603 | LINC00603 | -21.8kb | Anti-sense | Chr. 5 - 40052291 - 40053324 on NCBI Build GRCh38 | |
| Downstream | prostaglandin E receptor 4 | PTGER4 |
EP4, EP4R |
601.3kb | Anti-sense | Chr. 5 - 40679930 - 40740932 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map