Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1284 | MIR1284 |
MIRN1284, hsa-mir-1284, mir-1284 |
-25.8kb | Anti-sense | Chr. 3 - 71541970 - 71542089 on NCBI Build GRCh38 |
| Downstream | eukaryotic translation initiation factor 4E family member 3 | EIF4E3 |
eIF-4E3, eIF4E-3 |
108.2kb | Anti-sense | Chr. 3 - 71679289 - 71754831 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map