Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin 8 | PCDH8 |
ARCADLIN, PAPC |
-84.7kb | Anti-sense | Chr. 13 - 52843974 - 52848640 on NCBI Build GRCh38 |
| Downstream | olfactomedin 4 | OLFM4 |
GC1, GW112, OLM4, OlfD, UNQ362, bA209J19.1, hGC-1, hOLfD |
92.4kb | Sense | Chr. 13 - 53028741 - 53052061 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map