Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | receptor accessory protein 1 | REEP1 |
C2orf23, HMN5B, SPG31, Yip2a |
-98.6kb | Sense | Chr. 2 - 86213993 - 86338083 on NCBI Build GRCh38 |
| Downstream | charged multivesicular body protein 3 | CHMP3 |
CGI-149, NEDF, VPS24 |
62.7kb | Sense | Chr. 2 - 86503430 - 86563497 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map