Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724933 | LOC102724933 | -133.0kb | Sense | Chr. 12 - 93174366 - 93181832 on NCBI Build GRCh38 | |
| Downstream | nudix hydrolase 4 | NUDT4 |
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
62.2kb | Sense | Chr. 12 - 93377923 - 93403248 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map