Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoporin 35 | NUP35 |
MP-44, MP44, NP44, NUP53 |
-18.6kb | Anti-sense | Chr. 2 - 183117490 - 183161684 on NCBI Build GRCh38 |
| Downstream | microRNA 548ae-1 | MIR548AE1 | 1.2mb | Anti-sense | Chr. 2 - 184378975 - 184379044 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map