Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927269 | LOC101927269 | -47.1kb | Anti-sense | Chr. 7 - 80371517 - 80374439 on NCBI Build GRCh38 | |
| Downstream | G protein subunit alpha transducin 3 | GNAT3 | GDCA | 34.1kb | Anti-sense | Chr. 7 - 80457291 - 80512503 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map