Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane palmitoylated protein 6 | MPP6 |
PALS2, VAM-1, VAM1, p55T |
Sense | Chr. 7 - 24573346 - 24694086 on NCBI Build GRCh38 | |
| Downstream |
DFNA5, deafness associated tumor suppressor |
DFNA5 | ICERE-1 | 57.3kb | Anti-sense | Chr. 7 - 24698355 - 24758020 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map