Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724301 | LOC102724301 | -135.7kb | Sense | Chr. 11 - 119729583 - 119739623 on NCBI Build GRCh38 | |
| Downstream | tripartite motif containing 29 | TRIM29 | ATDC | 234.3kb | Anti-sense | Chr. 11 - 120111286 - 120138179 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map