Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506470 | LOC100506470 | -289.7kb | Sense | Chr. 20 - 61079064 - 61080179 on NCBI Build GRCh38 | |
| Downstream | microRNA 1257 | MIR1257 |
MIRN1257, hsa-mir-1257 |
582.7kb | Anti-sense | Chr. 20 - 61953546 - 61953662 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map