Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATP binding cassette subfamily B member 4 | ABCB4 |
ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3 |
-100.9kb | Sense | Chr. 7 - 87399461 - 87475864 on NCBI Build GRCh38 |
| Downstream | RUN domain containing 3B | RUNDC3B |
RPIB9, RPIP9 |
42.2kb | Anti-sense | Chr. 7 - 87627656 - 87832298 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map