Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mediator complex subunit 30 | MED30 |
MED30S, THRAP6, TRAP25 |
-428.3kb | Anti-sense | Chr. 8 - 117520546 - 117540262 on NCBI Build GRCh38 |
| Downstream | sterile alpha motif domain containing 12 | SAMD12 | 162.3kb | Sense | Chr. 8 - 118131828 - 118621995 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map