Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371907 | LOC105371907 | -74.5kb | Anti-sense | Chr. 17 - 77273784 - 77281897 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371905 | LOC105371905 | 13.6kb | Anti-sense | Chr. 17 - 77373730 - 77377341 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map