Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928371 | LOC101928371 | -33.3kb | Anti-sense | Chr. 2 - 88538720 - 88575610 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928403 | LOC101928403 | 18.1kb | Anti-sense | Chr. 2 - 88627829 - 88631825 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map