Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TAPT1 antisense RNA 1 (head to head) | TAPT1-AS1 | -623.4kb | Anti-sense | Chr. 4 - 16226663 - 16258187 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929123 | LOC101929123 | 288.0kb | Sense | Chr. 4 - 17171757 - 17186057 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map