Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoribosyl pyrophosphate synthetase 1 | PRPS1 |
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI |
-75.6kb | Anti-sense | Chr. X - 107628424 - 107651026 on NCBI Build GRCh38 |
| Downstream | midline 2 | MID2 |
FXY2, MRX101, RNF60, TRIM1 |
96.7kb | Anti-sense | Chr. X - 107820961 - 107931637 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map