Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 56B |
SNORD56B | RNU56B | -66.6kb | Sense | Chr. 14 - 71398337 - 71398407 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC145474 | LOC145474 | 21.3kb | Sense | Chr. 14 - 71487861 - 71489714 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map