Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EFCAB6 antisense RNA 1 | EFCAB6-AS1 | -175.4kb | Anti-sense | Chr. 22 - 43516254 - 43536605 on NCBI Build GRCh38 | |
| Downstream | sulfotransferase family 4A member 1 | SULT4A1 |
BR-STL-1, BRSTL1, DJ388M5.3, NST, SULTX3, hBR-STL-1 |
110.4kb | Sense | Chr. 22 - 43824507 - 43862498 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map