Assay Details
| Gene Symbol: LOC100507664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130016.1 | - | 1 | 1045 | 87 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BF329813.1 | - | 1 | Chr. 1 - 62608633 - 62608967 [-] on Build GRCh38 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dedicator of cytokinesis 7 | DOCK7 |
EIEE23, ZIR2 |
Anti-sense | Chr. 1 - 62454726 - 62688368 on NCBI Build GRCh38 | |
| Downstream | autophagy related 4C cysteine peptidase | ATG4C |
APG4-C, APG4C, AUTL1, AUTL3 |
174.5kb | Sense | Chr. 1 - 62784106 - 62865270 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100507664
Genomic Map