Assay Details
| Gene Symbol: LOC105369783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130110.1 | - | 1 - 2 | 123 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 26a-2 | MIR26A2 |
MIRN26A2, mir-26a-2 |
-12.4kb | Anti-sense | Chr. 12 - 57824609 - 57824692 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC283387 | LOC283387 | 26.6kb | Anti-sense | Chr. 12 - 57868056 - 57893819 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369783
Genomic Map