Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras protein specific guanine nucleotide releasing factor 2 | RASGRF2 |
GRF2, RAS-GRF2 |
-2.1kb | Anti-sense | Chr. 5 - 80960672 - 81230162 on NCBI Build GRCh38 |
| Downstream | CKMT2 antisense RNA 1 | CKMT2-AS1 | 3.7kb | Sense | Chr. 5 - 81237565 - 81301569 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map