Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CHRM3 antisense RNA 2 | CHRM3-AS2 | -142.3kb | Anti-sense | Chr. 1 - 239707126 - 239719119 on NCBI Build GRCh38 | |
| Downstream | CHRM3 antisense RNA 1 | CHRM3-AS1 | 33.9kb | Anti-sense | Chr. 1 - 239898019 - 239899872 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map