Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NLGN1 antisense RNA 1 | NLGN1-AS1 | -272.1kb | Anti-sense | Chr. 3 - 173910498 - 173920796 on NCBI Build GRCh38 | |
| Downstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | NAALADL2 | 246.2kb | Sense | Chr. 3 - 174439669 - 175809510 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map