Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spectrin repeat containing nuclear envelope protein 1 | SYNE1 |
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2 |
Sense | Chr. 6 - 152121684 - 152637399 on NCBI Build GRCh38 | |
| Downstream | SYNE1 antisense RNA 1 | SYNE1-AS1 | 4.6kb | Anti-sense | Chr. 6 - 152380530 - 152381713 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map