Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928361 | LOC101928361 | -73.4kb | Sense | Chr. 2 - 143598906 - 143608759 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928386 | LOC101928386 | 253.3kb | Sense | Chr. 2 - 143937067 - 143964156 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map