Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and SCAN domain containing 12 pseudogene 1 | ZSCAN12P1 |
ZNF187p1, ZNF305P1, ZNF96L1, ZNF96P1, ZSCAN12L1, dJ313I6.7 |
-20.2kb | Anti-sense | Chr. 6 - 28090807 - 28095715 on NCBI Build GRCh38 |
| Downstream | ZSCAN16 antisense RNA 1 | ZSCAN16-AS1 | 5.3kb | Sense | Chr. 6 - 28121795 - 28137316 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map