Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6129 | MIR6129 | hsa-mir-6129 | -15.6kb | Sense | Chr. 17 - 49288346 - 49288454 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102724596 | LOC102724596 | 55.5kb | Anti-sense | Chr. 17 - 49361165 - 49380094 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map