Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 8 open reading frame 31 | C8orf31 | Sense | Chr. 8 - 143038983 - 143054303 on NCBI Build GRCh38 | ||
| Downstream |
lymphocyte antigen 6 complex, locus L |
LY6L | 28.1kb | Sense | Chr. 8 - 143081045 - 143083467 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map