Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100130502 | LOC100130502 | -14.4kb | Anti-sense | Chr. 2 - 44921080 - 44923619 on NCBI Build GRCh38 | |
| Downstream | SIX3 antisense RNA 1 | SIX3-AS1 | 41.0 | Anti-sense | Chr. 2 - 44940154 - 44941878 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map