Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium voltage-gated channel subfamily H member 7 | KCNH7 |
ERG3, HERG3, Kv11.3 |
Sense | Chr. 2 - 162371407 - 162838747 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929570 | LOC101929570 | 45.4kb | Anti-sense | Chr. 2 - 162768936 - 162797972 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map