Assay Details
| Gene Symbol: LOC105370178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033984.2 | - | 1 | 48 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 428 | LINC00428 | -55.2kb | Anti-sense | Chr. 13 - 42842412 - 42872744 on NCBI Build GRCh38 | |
| Downstream | DnaJ heat shock protein family (Hsp40) member C15 | DNAJC15 |
DNAJD1, HSD18, MCJ |
94.6kb | Sense | Chr. 13 - 43023226 - 43109170 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105370178
Genomic Map