Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
surfactant associated 1, pseudogene |
SFTA1P | SFTPF | -129.8kb | Sense | Chr. 10 - 10784439 - 10794914 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 710 | LINC00710 | 7.1kb | Sense | Chr. 10 - 10934940 - 10952163 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map