Assay Details
| Gene Symbol: LOC105377371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037784.2 | - | 1 | 248 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 4 open reading frame 32 | C4orf32 | -40.5kb | Anti-sense | Chr. 4 - 112145397 - 112189081 on NCBI Build GRCh38 | |
| Downstream | adaptor related protein complex 1 associated regulatory protein | AP1AR |
2C18, C4orf16, GBAR, PRO0971, gamma-BAR |
141.0 | Anti-sense | Chr. 4 - 112231727 - 112273110 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105377371
Genomic Map