Assay Details
| Gene Symbol: ITGB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038209.2 | - | 1 - 2 | 48 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927811 | LOC101927811 | -215.0 | Anti-sense | Chr. 7 - 20328301 - 20331762 on NCBI Build GRCh38 | |
| Downstream | ATP binding cassette subfamily B member 5 | ABCB5 |
ABCB5alpha, ABCB5beta, EST422562 |
262.3kb | Sense | Chr. 7 - 20615622 - 20757014 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000222573.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000460204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000477859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000478974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | ENST00000537992.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006844.hg.1 | NM_002214.2 | Refseq |

ITGB8
Genomic Map