| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000420193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
ENST00000452247.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010654.hg.1 |
NR_038277.1 |
Refseq |