Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
lunapark, ER junction formation factor |
LNPK |
KIAA1715, LNP, LNP1, Ul, ulnaless |
Anti-sense | Chr. 2 - 175923862 - 176002901 on NCBI Build GRCh38 | |
| Downstream | even-skipped homeobox 2 | EVX2 | EVX-2 | 87.7kb | Anti-sense | Chr. 2 - 176080107 - 176083962 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map