Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928283 | LOC101928283 | -1.7mb | Anti-sense | Chr. 7 - 125184569 - 125379321 on NCBI Build GRCh38 | |
| Downstream | microRNA 592 | MIR592 |
MIRN592, hsa-mir-592, mir-592 |
26.9kb | Sense | Chr. 7 - 127058088 - 127058184 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map