Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 661 | MIR661 |
MIRN661, hsa-mir-661 |
-10.7kb | Sense | Chr. 8 - 143945191 - 143945279 on NCBI Build GRCh38 |
| Downstream | poly(ADP-ribose) polymerase family member 10 | PARP10 | ARTD10 | 19.5kb | Sense | Chr. 8 - 143977152 - 143986471 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map