Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cerebellar degeneration related protein 1 | CDR1 |
CDR, CDR34, CDR62A |
-5.3kb | Anti-sense | Chr. X - 140783260 - 140784558 on NCBI Build GRCh38 |
| Downstream | microRNA 320d-2 | MIR320D2 |
MIR320D-2, MIRN320D2, hsa-mir-320d-2, mir-320d-2 |
133.0kb | Anti-sense | Chr. X - 140926172 - 140926219 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map