Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate-glucosamine 3-sulfotransferase 3A1 | HS3ST3A1 |
3-OST-3A, 3OST3A1 |
-31.6kb | Sense | Chr. 17 - 13494032 - 13601942 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506974 | LOC100506974 | 141.9kb | Sense | Chr. 17 - 13790327 - 13898625 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map